3,3'-Diethyloxadicarbocyanine iodide (DODC) 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33094-03
H33094-06
H33094-MD
CAAAH33094-03EA
197.5
CAD
CAAAH33094-03
CAAAH33094-06
CAAAH33094-MD
3,3'-Diethyloxadicarbocyanine iodide (DODC) 96%
3,3'-Diethyloxadicarbocyanine iodide (DODC)
Purity: 96%
CAS No.: 14806-50-9
Molecular Formula: C23H23IN2O2
Formula Weight: 486.36
Melting Point: ca 215° dec.
MDL No.: MFCD00011953
Formula:
C₂₃H₂₃IN₂O₂ Melting Pt: ca 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011953 CAS Number: 14806-50-9 EINECS: 238-873-7 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...