Methyl-3-aminosalicylate 97%

Supplier: Thermo Scientific Chemicals
Warning

H33035-03 H33035-06
CAAAH33035-03EA 191.18 CAD
CAAAH33035-03 CAAAH33035-06
Methyl-3-aminosalicylate 97%
Methyl 3-aminosalicylate

CAS: 35748-34-6 MDL: MFCD09842614

Formula: C₈H₉NO₃
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD09842614
CAS Number: 35748-34-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR