2-Methyl-6-(trifluoromethyl)nicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33003-03
H33003-09
CAAAH33003-03EA
79.95
CAD
CAAAH33003-03
CAAAH33003-09
2-Methyl-6-(trifluoromethyl)nicotinic acid 97%
2-Methyl-6-(trifluoromethyl)nicotinic acid
CAS: 261635-93-2
MDL: MFCD01311990
Formula:
C₈H₆F₃NO₂ Melting Pt: 139…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311990 CAS Number: 261635-93-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...