4-Amino-2-methylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-o-toluic acid, 4-Amino-2-methylbenzoic Acid
H31657-03
H31657-06
CAAAH31657-03EA
25.75
CAD
CAAAH31657-03
CAAAH31657-06
4-Amino-2-methylbenzoic acid 98%
4-Amino-2-methylbenzoic acid
Purity: 98%
CAS No.: 2486-75-1
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Physical Form: Solid
MDL No.: MFCD06656133
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656133 CAS Number: 2486-75-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...