5-(4-Morpholinyl)-2-nitrophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

B24950-03 B24950-06
CAAAAB24950-03EA 82.32 CAD
CAAAAB24950-03 CAAAAB24950-06 CAAAAB24950-14
5-(4-Morpholinyl)-2-nitrophenol 97%
5-(4-Morpholinyl)-2-nitrophenol

CAS: 175135-19-0 MDL: MFCD00053057

Formula: C₁₀H₁₂N₂O₄
MW: 224.22 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00053057
CAS Number: 175135-19-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR