Magnesium carbonate hydroxide pentahydrate light 98% (by complexometric analysis)

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium carbonate basic pentahydrate, Magnesium hydroxide carbonate pentahydrate

A18070-0B A18070-0I A18070-30
CAAAA18070-0BEA 122.29 CAD
CAAAA18070-0B CAAAA18070-0I CAAAA18070-30
Magnesium carbonate hydroxide pentahydrate light 98% (by complexometric analysis)
Magnesium carbonate hydroxide pentahydrate

Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.


Purity: 98% CAS No.: 56378-72-4 Molecular Formula: C4H2Mg5O14.5H2O Formula Weight: 485.65 (395.58anhy) MDL No.: MFCD07371358


Caution: Stable under recommended storage conditions. Incompatible with oxidsing agents.

Formula: C₄H₂Mg₅O₁₄·5H₂O
MW: 485.65 g/mol (395.58 anhy)
Density: 2.160
Storage Temperature: Ambient
MDL Number: MFCD00149784
CAS Number: 56378-72-4
EINECS: 235-192-7

Order Now

Specification Test Results

Appearance (Colour) White
Form Powder
Complexometric Titration >97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR