Lead(II) acetate basic

Supplier: Thermo Scientific Chemicals
Danger

A16763-0E A16763-36
CAAAAA16763-0EEA 418.7 CAD
CAAAAA16763-0E CAAAAA16763-36
Lead(II) acetate basic
Lead (II) acetate basic

CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.72 Merck Reference: 14,5419 UN No.: 1616 MDL No.: MFCD00013041

Formula: (CH₃COO)₂Pb·2Pb(OH)₂
Storage Temperature: Ambient
MDL Number: MFCD00011154
CAS Number: 1335-32-6
EINECS: 215-630-3
UN: 1616
ADR: 6.1,III
Merck Index: 14,5419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR