Iron(III) hydroxide, α-phase ≥99%
Supplier: Thermo Scientific Chemicals
A16267-0B
A16267-30
CAAAA16267-0BEA
323.9
CAD
CAAAA16267-0B
CAAAA16267-30
Iron(III) hydroxide, α-phase ≥99%
Iron(III) hydroxide, alpha-phase
Purity: 99+%
CAS No.: 20344-49-4
Molecular Formula: HFeO2
Formula Weight: 88.85
Odor: Odorless
MDL No.: MFCD00064782
Formula:
FeO(OH) Density: 4.1 Storage Temperature: Ambient |
MDL Number:
MFCD00064782 CAS Number: 20344-49-4 EINECS: 243-746-4 Merck Index: 14,4025 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...