Micro Roll, Microfiber Wipes, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
149041
10769-328CS 638.84 CAD
10769-328
Micro Roll, Microfiber Wipes, Vileda Professional-FHP
Wipes Disposable Wipes and Towels Wipes for General Applications
A lightweight, true 100% microfiber wipe offering outstanding lint free cleaning performance with greater than 99.93% removal of bacteria.
  • 100% endless microfiber offers streak-free performance
  • Lint-free cleaning performance with greater than 99.93% bacteria removal
  • One step cleaning with no additional workload
  • Better cleaning results in less time
  • Suitable for bucket or spray cleaning methods
This semi-disposable cloth features one-step cleaning and streak free performance. Achieve better cleaning results in less time, and these wipes are suitable for bucket or spray cleaning methods as well.
Ordering information: Wipes are packaged 200 wipes per roll with 4 rolls per case. Wipes are 50 gm per sheet.