Protein A Sepharose™ CL-4B Affinity Chromatography Media, Cytiva

Supplier: Cytiva

17-0963-02 17-0780-01 17-0963-03
CA95055-962LEA 51479.87 CAD
CA95055-962L CA95016-992L CA95017-044L
Protein A Sepharose™ CL-4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins

Protein A binds to the Fc region of immunoglobulins through interactions with the heavy chain.


  • For affinity chromatography
  • For immunoglobin purification and fractionation, immunoprecipitation procedures and polyclonal antibody purification
  • Strong binding to Fc region of IgG
  • High capacity - 20 mg human IgG/ml
  • Cyanogen bromide activation coupling method
  • Stable in all commonly used aqueous buffers, 6 M guanidine-HCl, 8 M urea
  • Freeze-dried powder or preswollen in 20% ethanol
  • Negligible volume variation due to changes in pH or ionic strength


The binding of protein A has been well documented for IgG from a variety of mammalian species and for some IgM and IgA as well. Protein A Sepharose CL-4B has been used as a powerful tool to isolate and purify classes, subclasses and fragments of immunoglobulins from biological fluids and from cell culture media. Since only the Fc region is involved in binding, the Fab region is available for binding antigen.

Order Now

SPECIFICATIONS

Ligand Native Protein A
Environmentally Preferable
Matrix Cross-linked agarose, 4%
Flow rate <150 cm/h
pH range 3 - 9
Binding capacity ≈ 20 mg human lgG/mL medium
Storage temperature 2 - 8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR