IMAC Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
17-0921-08 17092107
CA95017-034EA 2708.94 CAD
CA95017-034 CA97067-852
IMAC Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.
  • For affinity chromatography
  • Uncharged IMAC resin allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Convenient purification of histidine-tagged proteins, also when Ni2+ is not the best choice of metal ion
  • Stable in commonly used aqueous buffers 0.01 M HCl, 1 M NaOH, 70% acetic acid, 2% SDS. 30% 2-propanol
IMAC Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification. As a BioProcess medium, IMAC Sepharose 6 Fast Flow fulfills industrial demands for security of supply, robust performance and regulatory support. The coupling chemistry and the novel ligand is designed to give minimal ligand leakage while providing high capacity.
IMAC Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, IMAC Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
IMAC Sepharose 6 Fast Flow is supplied free of metal ions, allowing the user to charge it with the most appropriate metal ion for purification of a target protein
Ordering information: Available in the convenient and time-saving prepacked HiTrap and HiPrep formats.