Whatman™ Cellulose Blotting Papers, Grade GB005, Whatman products (Cytiva)
Supplier: Cytiva
10426972 10426981 10426994 10426972 10426981 10426994
89026-874PK 185.35 CAD
89026-874 89026-876 89026-878 CA89026-874 CA89026-876 CA89026-878
Whatman™ Cellulose Blotting Papers, Grade GB005, Whatman products (Cytiva)
Blotting Papers
Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind
Ensures that no contamination will occur during the transfer steps. Manufactured and tested specifically for chromatographic and blotting techniques. This ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting. Whatman 3MM Chr is considered the industry standard for blotting procedures. Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes. Allows "out-of-More Product Information
Order Now
SPECIFICATIONS
Grade GB005
Environmentally Preferable