Superdex 200 Increase Small-Scale SEC Columns, Cytiva
Supplier: Cytiva
28990944 28990945 28990946
CA11029-390EA 6928.86 CAD
CA11029-390 CA11029-392 CA11029-394
Superdex 200 Increase Small-Scale SEC Columns, Cytiva
Chromatography Columns
Superdex 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10000 to 600000.
  • Enhanced performance: Improved resolution and runtime compared to predecessor Superdex 200
  • Very high resolution: Small bead size and narrow particle size distribution provide high resolution, for high protein purity
  • High flow rates: Rigid beads give excellent pressure/flow properties
Superdex 200 Increase is a next-generation, dextran-More Product Information