Amersham™ Protran® Supported Nitrocellulose Western Blotting Membranes, Cytiva
Supplier: Cytiva
10600015 10600016 10600017 10600018 10600019 10600020 10600120 10600037 10600119 10600051 10600053
CA10061-150PK 1026.97 CAD
CA10061-150 CA10061-152 CA10061-154 CA10061-202 CA10061-204 CA10061-206 CA10061-298 CA10061-326 CA10061-338 CA10061-416 CA10061-420
Amersham™ Protran® Supported Nitrocellulose Western Blotting Membranes, Cytiva
Transfer Membranes
Amersham™ Protran® Supported is a robust protein blotting membrane, with 0,2 µm pore size, made of reinforced nitrocellulose for multiple reprobings.
  • Optimal for applications when stripping and reprobing is required
  • Versatile, robust, and excellent signal-to-noise ratios
  • High binding capacity
  • Protran® Supported 0,2 NC membrane recommended for detection of small proteins and peptides (
  • Protran® Supported 0,45 µm membrane suitable for proteins of a wide range of molecular weights
Amersham™ Protran® Supported offers excellent sensitivity, resolution, andMore Product Information