Uni-Core Punch, Qiagen
Supplier: Qiagen
WB100028 WB100029 WB100039 WB100040 WB100073 WB100074 WB100076 WB100078 WB100082 WB100074 WB100028 WB100029 WB100039
14230-568PK 232.34 CAD
14230-568 14230-570 89022-356 89022-358 CA30621-462 CA30621-464 CA30621-468 CA30621-472 CA30621-480 30621-464 CA14230-568 CA14230-570 CA89022-356
Uni-Core Punch, Qiagen
Samplers Solid Samplers
Uni-Core Punch is a disposable punch with a polished steel tip that is case-hardened and can be sterilized
  • Designed for use with FTA Cards, FTA Elute Cards, and EasiCollect devices
  • Capable of up to 500 punches
  • Multiple diameters ranging from 0.35–12.0 mm in plastic housing for sample punching versatility
  • Stainless steel cutting edges enable manual coring, retrieval, and controlled ejection of the FTA punch
  • Disposable to minimize contamination and carryover between samples when a cleaning procedure is followed
It is housed in a plastic handle and disposable for contamination control. Manual punching is preferred for laboratories with low sample volumes punching 1 to 50 samples per day.