Tuerk solution
Supplier: AZER SCIENTIFIC INC
Synonyms:
Acetic acid gentian violet solution
, Turk solution
ES930
ES932
ES931
89423-628EA
56.6
CAD
89423-628
89423-632
89423-630
Tuerk solution
Tuerk solution
Turk's blood diluting fluid has a stain (gentian violet) plus 6% acetic acid and is ideal for manual leukocyte counting.
- Ideal for Manual Leukocyte Counting
- Six Percent Acetic Acid
- Gentian Violet Stain
Stains the nuclei of the white blood cells making them easier to see and count.