Reaction Head for Pilot Plant, Ace Glass
Supplier: Ace Glass
6530-36 6530-38 6530-39 6530-40 6472-22 6472-23 6530-31 6530-32 6530-33 6530-35 6530-37 6530-45 6530-50 6530-60 6530-70 6530-30 6530-34 6530-85 6530-28 6530-46 6530-75
71001-534EA 3479.38 CAD
71001-534 71001-536 71001-538 71001-540 89054-652 89054-654 89055-368 89055-370 89055-372 89055-374 89055-376 89055-386 89055-394 89055-402 89084-062 89084-088 89084-090 89427-926 10767-300 10767-302 76107-012
Reaction Head for Pilot Plant, Ace Glass
Lids Reaction Vessel Lids
Borosilicate glass domed heads with reinforced standard taper joints for pilot plant reactors
Some heads are available with angled addition ports, for ease in adding materials to the reactor.
Heads have either DURAN® or KF flanges, standard taper joint center necks (45/50) and side necks.
Ordering information: Head does not include addition port cap, O-ring, or clamp, they must be ordered separately.