Voltage Converter, Step Up & Down Transformer, Ace Glass Incorporated
Supplier: Ace Glass
7834-17
10767-314EA 1189.62 CAD
10767-314
Voltage Converter, Step Up & Down Transformer, Ace Glass Incorporated
Power Supplies
2000 watt voltage converter can be used as a step up transformer in 110/120 volt countries or as a step down transformer in 220/230/240 volt countries.
It will convert voltages of 220-240 to 110-120 volts and will also convert voltages from 110-220 to 220-240 volts. Operates in either 50 or 60Hz, but will not change frequency.
Fuse protected and supplied with two spare fuses. Features a 220 volt universal output plug which will accept most International plugs.
Certifications: CE certified for continuous use.