PTFE Tube Compression Union, Ace Glass Incorporated
Supplier: Ace Glass
12721-04 12721-06 12721-08 12721-10 12721-12 12721-14 12721-20 12721-22 12721-24 12721-28
10767-430EA 169.48 CAD
10767-430 10767-432 10767-434 10767-436 10767-438 10767-440 10767-442 10767-444 10767-446 10767-448 10767-450 10767-452 10767-454 10767-456 10767-458
PTFE Tube Compression Union, Ace Glass Incorporated
Fittings and Connectors
The PTFE tube compression unions have body and ferrules made of virgin PTFE
Performance at ambient room conditions: 120psi for 1/16" fittings linear decreasing to 80psi for 3/4" fittings and 60psi for fittings larger than 3/4".
Performance at elevated temperatures up to 85C/185F: 90psi for 1/16" fittings linear decreasing to 60psi for 3/4" fittings and 40psi for fittings larger than 3/4".
The nut and ferrule-gripper are made of PVDF.
Order Now
SPECIFICATIONS
Environmentally Preferable