Vertical Adapter with Drip Tip, Joint to Tube Compression, Ace Glass Incorporated
Supplier: Ace Glass
12736-01 12736-03 12736-07 12736-09 12736-11 12736-13 12736-15 12736-17 12736-19 12736-21 12736-23 12736-25 12736-27
10767-480EA 122.98 CAD
10767-480 10767-482 10767-484 10767-486 10767-488 10767-490 10767-492 10767-494 10767-496 10767-836 10767-838 10767-840 10767-842
Vertical Adapter with Drip Tip, Joint to Tube Compression, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Vertical adapter featuring a drip tip.
  • Vertical sidearm
  • [ST] joint
  • Drip tip
Connects standard taper joint to tubing.