Right Angle Tee Joint to Tube Compression Adapter with Drip Tip, Ace Glass Incorporated
Supplier: Ace Glass
12739-23 12739-25 12739-27 12739-01 12739-03 12739-07 12739-09 12739-11 12739-13 12739-15 12739-17 12739-19 12739-21
10767-512EA 291.69 CAD
10767-512 10767-514 10767-516 10768-058 10768-060 10768-062 10768-064 10768-066 10768-068 10768-070 10768-072 10768-074 10768-076
Right Angle Tee Joint to Tube Compression Adapter with Drip Tip, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These glass standard taper joint adapters feature a drip tip and right angle tee ground glass tube sidearms.
  • Glass [ST] joint
  • Drip tip
  • Right angle tee sidearms
The sidearms are used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.