Jacketed Kilo Scale Reactor, Ace Glass Incorporated
Supplier: Ace Glass
12845-05 12845-06 12845-07 12845-02 12845-04 12845-03 12845-09 12845-11 12845-13 12845-15 12845-17 12845-19
10767-200EA 44354.79 CAD
10767-200 10767-202 10767-204 10767-518 10767-860 10768-078 10768-080 10768-082 10768-084 10768-086 10768-088 10768-090
Jacketed Kilo Scale Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
A jacketed reactor base system that is designed for maximum diversity and ease of use
Each base system comes with the basic starting components required. The motor and accessories should be purchased separately, based upon your specific needs.
Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom-designed parts.
Ordering information: Base reactor system includes universal stand with motor mount, jacketed flask, multi-neck head, O-ring/gasket and quick-release clamp for head, cap, O-ring, and quick-More Product Information