Jacketed Cylindrical Flask with Beaded Pipe Inlet and Outlet, Ace Glass
Supplier: Ace Glass
12850-04 12850-14 12850-02 12850-16 12850-11 12850-10 12850-08 12850-06 12850-15 12850-12
76214-494EA 26973.54 CAD
76214-494 10768-120 10768-122 10768-124 10768-114 10768-126 10768-116 10767-208 10768-118 10767-206
Jacketed Cylindrical Flask with Beaded Pipe Inlet and Outlet, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
A jacketed cylindrical flush seal flask with beaded pipe inlet and outlets.
  • Flush seal valve is removable for easy cleaning
The flush seal valve incorporates a 1" beaded pipe angled side arm for maximum unobstructed drainage. The maximum temperature range is -60 to +200°C. The maximum jacket pressure is 5 PSI. The flask is graduated on both sides, which allows for either left or right orientation. 10 to 20L flasks feature Duran style o-ring groove flanges, and the 30 to 150L flasks feature KF Plane flat flanges.