Digivac 22W LCD Vacuum Transmitter, Ace Glass Incorporated
Supplier: Ace Glass
14302-01
10767-868EA 2366.84 CAD
10767-868
Digivac 22W LCD Vacuum Transmitter, Ace Glass Incorporated
Vacuum Handling Systems
This small compact versatile vacuum gauge can fit almost anywhere
The vacuum gauge has a built in RS232 port for data download to a PC, and a 5 vdc output with a single set-point for output to PLC's or chart recorders. The range is 1 milliTorr to 760 Torr. The vacuum gauge power is 100-230V 50/60Hz.
The vacuum gauge uses a standard, replaceable, vacuum gauge tube with 1/8"MNPT threads. It can be adapted to fit onto any schlenk line to give accurate and recordable data readings.
Certifications: Factory calibrated to NIST traceable standards. CE rating.