Axygen® 96-Well Half Skirt Automation Compatible PCR Microplate, Corning
Supplier: Corning
PCR-96-HS-AC-C PCR-96-HS-AC-C-PBC PCR-96-HS-AC-C-3BC
10015-636CS 650.11 CAD
10015-636 CA10015-636 75784-632 75786-266
Axygen® 96-Well Half Skirt Automation Compatible PCR Microplate, Corning
Microplates PCR Plates
This automation-compatible, polypropylene PCR microplate features a rigid frame with a small split that sustains heat transferring during and post thermal cycling without distorting its shape. The plate also features raised rims around the wells to provide tight sealing to minimize heat evaporation.
  • Remains flat during and post thermal cycling
  • Raised well rims help reduce heat evaporation
  • Clear wells, visible grid for easy sample loading and tracking
  • Ultra-thin and consistent wall thickness for easy and consistent heat transfer
  • Single notch corner
  • SBS
More Product Information