Individual Slotted Killogram Weights
Supplier: Wards
PH0264B2 PH0264B3 PH0264B4
470219-776EA 40.9 CAD
470219-776 470219-778 470219-780
Individual Slotted Killogram Weights
Weights Calibration Weights
Individual cast iron slotted weights.
  • Available individually
  • Stackable
  • Made from solid cast iron
  • Finished with a black enamel
  • Stamped with weight value
Slotted iron masses are suitable for use with sonometers or other applications, where a large load is required. The slotted masses can be hung on a hanger.
Ordering information: Hanger ordered separately.