Ward's® Comprehensive Periodic Table
Supplier: Avantor
470165-562CS 924.49 CAD
470165-562
Ward's® Comprehensive Periodic Table
Charts
This chart displays detailed and up-to-date information.
  • Double-Sided Formats Available
  • Three-Color Chart
  • Laminated Student Fill-In Form
  • Radioisotope Table
  • Subatomic Particle Table
Ward's comprehensive periodic table provides students with detailed and up-to-date information. In addition to elemental data, side one includes a table of radioisotopes with mass number, half-life, and decay particles. Side two includes a table of subatomic particles. These three-color charts come in a variety of sizes to meet all your teaching needs. Information updated in December 2016.
The student form is also printed on both sides and contains symbols, column markers, row numbers, and chart outlines. The laminated versions are faced in vinyl on both sides, so writing on the tables with a wet-erase pen or grease pencil is a snap.