Ward's® Advanced Periodic Table
Supplier: MERCURY PRINT PRODUCTIONS SE
402_U ADVFREN
470217-480PK 121.09 CAD
470217-480 470334-204
Ward's® Advanced Periodic Table
Charts
Ward's Advanced Periodic Table is available in a variety of sizes to meet all teaching needs.
  • Most Thorough Periodic Table Poster Available
  • Colored Poster
  • Double-Sided Formats Available
  • Information Organized in Simple, Logical Format
Our most complete periodic table, the chart has a highly visible element symbol, name, and atomic number. The nine important elemental properties–atomic weight, density, melting point, boiling point, atomic radius, electronegativity, first ionization energy, oxidation states, and electron configuration–are organized in a logical and easy-to-read format. Information updated December 2016.
The back side of the 8.5 x 11" and 11 x 17" versions display selected polyatomic ions, solubility rules, prefixes, unit abbreviations, useful equations, constants, and periodic trends. Teachers can hang the larger tables on a classroom wall to reference while teaching.