Diskcover-47 Extraction Disk Holder, Restek
Supplier: Restek
24020 24021 24025 24017
RK24020EA 380.4 CAD
RK24020 RK24021 RK24025 RK24017
Diskcover-47 Extraction Disk Holder, Restek
Chromatography Supplies Chromatography Spare Parts
The Diskcover-47 Extraction Disk Holder is compatible with most vacuum manifold systems that accept 1/8" male luer fittings.
  • Compatible with many systems
  • Easy automation of sample introduction
Samples can be automatically introduced via 1/8" PTFE tubing, or from the optional Diskcover-47 reservoir.