Screw Top Pathology Containers, Azer Scientific
Supplier: Azer Scientific Inc.
ES60120 ES60180 ES60240 ES60480 ES61000
10159-040CS 168.83 CAD
10159-040 10159-042 10159-044 10159-048 10159-050
Screw Top Pathology Containers, Azer Scientific
Sample Containers General Purpose Containers
Manufactured from high-quality, clarified polypropylene and resistant to histology reagents.
  • High-Quality, High-Density Polyethylene
  • Screw Top Lid
  • Leak-Resistant Closure
  • Histology Reagent Resistant
The screw top lid provides a leak-resistant closure to help protect the specimen and increase safety for lab personnel.
Order Now
SPECIFICATIONS
Environmentally Preferable