EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98%, Calbiochem®

Supplier: MilliporeSigma
Warning

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

Packaged under inert gas.

341006-25GM 341006-5GM
CA80051-566EA 770.42 CAD
CA80051-566 CA80051-568
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98%, Calbiochem®
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

EDAC HCl is a water-soluble derivative of carbodiimide useful for conjugating haptens to proteins and polypeptides.


Used to modify NMDA receptors and as a condensing agent in peptide synthesis.


Special Instructions: Unstable in solution. Reconstitute just prior to use.

Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Melting Pt: 112…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% by Titration
Form White solid
Molecular Formula C₈H₁₇N₃ · HCl
Molecular Weight 191.7
Toxicity Irritant
Storage -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR