Stable 'Z'® Stage Adapters, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
40319261914N 40319261915O 40319261919S 4031323
10199-842EA 2579.54 CAD
10199-842 10199-844 10199-846 10199-850
Stable 'Z'® Stage Adapters, Bioptechs Inc.®
Fittings and Connectors
The Stable "Z"® Nikon Stage Adapter is a 108 mm round glass plate with an electronically heated metal plate in its center to fit a Nikon microscope stage where biological specimens are placed for observation.
  • Minimal mass of heating component
  • Specimen is coplanar with support surface to eliminate Z axis drift
  • Minimizes heat transfer to the stage
  • Adjustable specimen plane to accommodate various dishes
The Stable "Z"® Olympus Stage Adapter is a 110 mm round glass plate with an electronically heated metal plate in its center that fits an Olympus microscope stage where biological specimens are placed for observation. The Stable "Z"® SBS Stage Adapter is a 127.76×85.48 mm glass plate with an electronically heated metal plate in its center that accepts 35 mm dishes and fits microscope stages adapted to hold 96-Well SLAS/SBS plates. The Stable "Z"® Lab-Tek™ Warming Stage Adapter is a 127.76×5.48 mm glass plate with an electronically heated metal plate in its center that accepts Lab-Tek™ chamber slides and fits microscope stages adapted for 96-Well SLAS/SBS plates. All adapters offer the ability to not induce Z axis drift to the specimen.
Certifications: CE certified.
Delivery information: Includes a reference temperature probe.