ICD Heated Lid, Bioptechs Inc.®
Supplier: Bioptechs
19030812
10199-872EA 1899.29 CAD
10199-872
ICD Heated Lid, Bioptechs Inc.®
Seals Gas Stops
This O-Ring sealed, stainless steel insert is equipped with a heated glass window to prevent condensation and atmospherically seal an ICD.
  • Comes with CO2 port
  • Prevents outside contamination
  • Use with interchangeable coverglass dish
Lid has a gas flow port and allows trans-illumination on an inverted microscope.
Certifications: CE certified.