Glass Culture Cylinders and Cylinder Starter Set, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
0703031919 07030301 07030302 07030304 07030306 07030308 07030310 07030312 07030314
10199-820KT 546.54 CAD
10199-820 10199-822 10199-824 10199-826 10199-828 10199-830 10199-832 10199-834 10199-836
Glass Culture Cylinders and Cylinder Starter Set, Bioptechs Inc.®
Culture Dishes Cell Culture Dishes
An assortment of borosilicate glass cylinders, all measuring 5 mm high, each with an optically flat polished end to eliminate the need for grease when plating cells.
  • Restrict and concentrate the growth of cells
  • Heavier than plastic cloning rings
  • Eliminate floating
  • Seal without grease
  • No contamination
This facilitates positioning for observation and improves plating time. Culture cylinders are reusable and autoclaveable.
Certifications: CE certified.
Ordering information: Starter Set includes 2, 4, 6, 8, and 10 mm sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable