Objective Thermal Spacers, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
152019R 152019 150827
10199-806EA 370.54 CAD
10199-806 10199-808 10199-810
Objective Thermal Spacers, Bioptechs Inc.®
Fittings and Connectors
These 9 mm thick internally and externally threaded polymer rings are to be placed between an objective and nosepiece to inhibit thermal conductivity.
It is recommended to add spacers to other objectives on the nosepiece for parfocality. Available in three formats: for RMS, 0.8"×36 tpi, 25 mm for 25×0.75 mm objectives, or 27 mm for 27×0.75 mm objectives.
Certifications: CE certified.