SpectraMax® i3x Multi-Mode Detection Platform, Molecular Devices
Supplier: Molecular Devices
SpectraMax®
I3X I3X
CA10014-924EA 129068.04 CAD
CA10014-924 10014-924
SpectraMax® i3x Multi-Mode Detection Platform, Molecular Devices
Microplate Readers
The SpectraMax® i3x Multi-Mode Detection Platform measures spectral-based absorbance, fluorescence, and luminescence with the added functionality of modular upgrades for western blot, imaging, and fast kinetics with injectors.
  • Modular upgrades
  • Expanded dynamic range
  • Spectral Fusion™ illumination
  • Power consumption <200 watts
The unit allows users to unravel the mysteries of science by exploring cellular pathways and protein activation and expression in one system.
The SpectraMax® i3x offers an expanded dynamic range. It is engineered for performance with Spectral Fusion™ illumination for increased sensitivity across the entire excitation range. The SpectraMax® i3x also features a cooled photomultiplier tube (PMT) for improved detection in extremely low light. These features enable users to generate more data points without the need to dilute.
The unit provides a variety of user-upgradeable applications. User-exchangeable detection modules expand the system’s detection capabilities to include Time-Resolved Fluorescence, HTRF, Fast Kinetics with Injectors, and Western Blot detection.
The SpectraMax® i3x Multi-Mode Detection Platform is one complete solution. With available options such as the SpectraMax® MiniMax™ 300 Imaging Cytometer, ScanLater™ Western Blot cartridge, reagents optimized for high performance, and the industry-leading data acquisition and analysis tool SoftMax Pro, the SpectraMax® i3x Multi-Mode Detection Platform is the total solution for all research needs. Let Molecular Devices and the SpectraMax® i3x Multi-Mode Detection Platform power the next landmark discovery.
Ordering information: Base system includes: Spectral Absorbance, Fluorescence, and Luminescence detection and 3 top read cartridge positions for user-upgradable detection cartridges; gas inlet for environmental control-capability; spectral fusion illumination; SoftMax Pro software; integrated shaking and temperature control, power cable, USB cable, compatible with 6-, 12-, 24-, 48-, 96-, and 383- and/or 1536-well plates. Compatible with SpectraMax® Imaging Cytometer option and/or SpectraDrop™ Micro-Volume microplate. Supplied with one-year warranty covering parts and labor.