FCS3® Non-Heating Chamber Set, Bioptechs®
Supplier: Bioptechs
210603193NH
10200-112KT 7869.63 CAD
10200-112
FCS3® Non-Heating Chamber Set, Bioptechs®
Flow Cells
Compatible, microscopic flow cell with a user definable specimen area for upright microscopes.
  • Unparalleled, uniform temperature control across entire specimen area
  • Near laminar flow perfusion
  • User definable flow characteristics
  • Compatible with all modes of microscopy including high NA/high magnification uses
  • Over 20 years of being the foremost chamber in micro-environmental control
  • Ideal for both flow and static applications
  • A parallel plate, highly N.A.
Certifications: CE certified.
Ordering information: Order includes Chamber only.