FCS3® Closed Chamber System Starter Set, Bioptechs Inc.®
Supplier: Bioptechs
210603193
10200-110KT 17488.02 CAD
10200-110
FCS3® Closed Chamber System Starter Set, Bioptechs Inc.®
Flow Cells
The FCS3® Closed Chamber System is a closed system, parallel plate, micro-environmental chamber for upright microscopes.
  • Unparalleled, uniform temperature control across entire specimen area
  • Near laminar flow perfusion
  • User definable flow characteristics
  • Systems for upright microscopes
  • Compatible with all modes of microscopy including high NA/high magnification uses
  • Over 20 years of being the foremost chamber in micro-environmental control
  • Ideal for both flow and static applications
The FCS3® provides the user with all of the functionality of the popular FCS2®, but is redesigned for upright microscopes. The system is designed specifically for the demands of today's live-cell imaging requirements for upright microscopes. It offers limitless flow characteristics because its flow geometry can easily be customized by the user. It can provide low shear near laminar flow as well as high shear narrow directed flow and anything in between. The most important feature of the FCS3®, other than unequalled temperature uniformity, is that there is a precise directed flow of media over the cells. The FCS3® provides an optical imaging cavity allowing the user to precisely define the optimum flow characteristics necessary for the experiment.
Certifications: CE certified.
Ordering information: Starter Set includes FCS3® Chamber, Controller, five Microaqueduct Slides, one hundred #1.5 thickness, 40mm German Desag263 Coverslips, and 30-Piece Gasket Set.