FCS2® Chamber, Bioptechs
Supplier: BIOPTECHS INC.
060319203
10200-006EA 7213.84 CAD
10200-006
FCS2® Chamber, Bioptechs
Flow Cells
A parallel plate, high N.A
  • Unparalleled, uniform temperature control across entire specimen area
  • Near laminar flow perfusion
  • User definable flow characteristics
  • Compatible with all modes of microscopy including high NA/high magnification uses
  • Over 20 years of being the foremost chamber in micro-environmental control
  • Ideal for both flow and static applications
compatible, uniformly temperature controlled microscopic flow cell with a user definable specimen area.
Certifications: CE certified.
Ordering information: Chamber only, requires FCS2® Controller.