FCS2® Single Tubing Sets, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
060319192016 060319192016S 060319031C31 060319020C20 060319015S15
10200-098PK 408.27 CAD
10200-098 10200-100 10200-102 10200-104 10200-106
FCS2® Single Tubing Sets, Bioptechs Inc.®
Tubing Transfer Tubing
FCS2® Single Tubing Sets include a package of four single C-Flex or Silicone tubings with molded ends to fit the Micro-Perfusion Pump
Tubing is available in a variety of thicknesses and material formats. The 0.062" Int. Ø tubing (C-Flex or Silicone) delivers flow rates of 125 to 1250 µl/min. from a low-flow pump, 10 to 100 µl/min. for a, ultra low-flow pump, and 0.9 to 9 ml/min. for a high-flow pump. The 0.031" Int. Ø tubing (C-Flex only) delivers flow rates of 35 to 350 µl/min. for low-flow pumps, 3 to 30 µl/min. for ultra low-flow pumps, and 0.3 to 2.8 ml/min. for high flow pumps. The 0.020" Int. Ø tubing (C-Flex only) delivers flow rates of 18 to 180 µl/min. for low-flow pumps, 1.6 to 16 µl/min. for ultra low-flow pumps, and 0.15 to 1.5 ml/min. for high-flow pumps. The 0.015” Int. Ø tubing (Silicone only) delivers flow rates of 8 to 83 µl/min. for low-flow pumps, 0.8 to 7.5 µl/min. for ultra low-flow pumps, and 0.07 to 0.7 ml/min. for high-flow pumps.
Certifications: CE certified.