FCS2® Closed Chamber System Starter Set, Bioptechs Inc.®
Supplier: Bioptechs
0603192
10200-004KT 17488.02 CAD
10200-004
FCS2® Closed Chamber System Starter Set, Bioptechs Inc.®
Flow Cells
The Focht Chamber System 2 (FCS2®) is a closed system, live-cell micro-observation chamber that offers several advantages over other chambers
  • Unparalleled, uniform temperature control across entire specimen area
  • Near laminar flow perfusion
  • User definable flow characteristics
  • System for inverted microscopes
  • Compatible with all modes of microscopy including high NA/high magnification uses
  • Over 20 years of being the foremost chamber in micro-environmental control
  • Ideal for both flow and static applications
In addition to its unique perfusion and thermal control systems, it is fully compatible with all modes of microscopy. It is also the only chamber to combine high-volume laminar flow perfusion rates with Koehler illumination and precise temperature control without an air curtain. The most important feature of the FCS2®, other than unequalled temperature uniformity, is the precise, directed flow of media over the cells. The system provides an optical imaging cavity allowing users to precisely define the optimum flow characteristics necessary for the experiment.
Certifications: CE certified.
Ordering information: Starter Set includes, FCS2® Chamber, Controller, five Micro aqueduct Slides, one hundred #1.5 thickness, 40mm German Desag263 Coverslips, and 30-piece Gasket Set.