Dithiothreitol (DTT, Cleland's reagent), SPUTOLYSIN®

Supplier: MilliporeSigma

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

560000-1SET 560000-10ML
CA80058-634EA 969.89 CAD
CA80058-634 CA80602-670
Dithiothreitol (DTT, Cleland's reagent), SPUTOLYSIN®
Dithiothreitol (DTT, Cleland's reagent)

Sputolysin reagent is a concentrate of dithiothreitol (DTT) in phosphate buffer intended for isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi and yeasts from sputum.


  • Epethelial cell isolation
  • Concentrate of dithiothreitol (DTT) in phosphate buffer
  • Isolation from sputum
  • pH 6.5 - 7.5


Each vial is sufficient to make 100 ml of working solution.

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004877
CAS Number: 3483-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR