Perfusion Pump Double Tubing Sets, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
0420042016 0420042016S 042004031C31 0420020C20
10199-992PK 406.34 CAD
10199-992 10199-994 10199-996 10199-998
Perfusion Pump Double Tubing Sets, Bioptechs Inc.®
Tubing Transfer Tubing
Double Tube Pump Sets are available in a variety of inner diameter thicknesses with manifold ends to fit the Bioptechs Micro-Perfusion Pumps.
The 0.062 Int. Ø sets (available in C-Flex or Silicone) deliver flow rates of 125 to 1250 µl/min. from a low flow pump, 10 to 100 µl/min. from an ultra-low flow pump, and 0.9 to 9 ml/min. from a high flow pump. The 0.031" Int. Ø sets (C-Flex only) deliver flow rates of 35 to 350 µl/min. from a regular pump, 3 to 30 µl/min. from a low flow pump, and 0.3 to 2.8 ml/min. from a high flow pump. The 0.020" Int. Ø sets (C-Flex only) deliver flow rates of 18 to 180 µl/min. from a regular pump, 1.6 to 16 µl/min. from a low flow pump, and 0.15 to 1.5 ml/min. from a high flow pump. The 0.062" (¹⁄₁₆") C-Flex tubing is also available in a 50' roll. All tubing is USP Class VI certified for maximum biocompatibility when using perfusion pumps.
Certifications: CE certified.
Ordering information: Tubing sets are designed for use with Bioptechs Micro-Perfusion Pumps.