Blank Gaskets, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
1907100 1907250 1907500 1907750
10200-014PK 109.34 CAD
10200-014 10200-016 10200-018 10200-068
Blank Gaskets, Bioptechs Inc.®
Seals Gaskets
These flat, 40 mm diameter silicone sheets are available in varied thicknesses, and can be modified by the user to suit specific protocols.
  • Unlimited options for internal geometry
  • Creates user definable flow characteristics
  • Enables near laminar flow
Ordering information: Each order includes 5 gaskets.