Glass Coverslips, 40mm, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
4013130319 40131303192 40131303193
10200-060PK 692.89 CAD
10200-060 10200-062 10200-064
Glass Coverslips, 40mm, Bioptechs Inc.®
Cover Glasses
These 40mm German Desag 263 borosilicate glass coverslips are ideal for imaging applications
  • Ideal for imaging
  • Desag 263 low-alkaline glass for cell adhesion
Available in packages of 200, 500, or 1000.