Delta T® TPG Non-Heated Culture Dishes, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
0420040500 0420041500C 0420041500B
10199-958PK 117.95 CAD
10199-958 10199-960 10199-962
Delta T® TPG Non-Heated Culture Dishes, Bioptechs Inc.®
Culture Dishes Cell Culture Dishes
These 35mm diameter dishes are available in multiple colors and thicknesses to accommodate a variety of applications
Easily adapted to a wide variety of specimen types from monolayered adherent cells to brain slice and tissue preps
Plate, incubate and observe without the need to transfer your cells
Coverglass bottom for optimum optical compatibility
Clear dishes are available with 0.5mm or 0.17mm thick glass bottoms, and are to be used for low N.A. applications without the ITO heating coating. Black dishes feature a 0.17mm thick clear Desag 263 German glass bottom with a black perimeter used for high N.A. imaging without the ITO heating coating. Black is offered for TIRF and fluorescence imaging applications.
Certifications: CE certified.
Ordering information: Each package includes 10 dishes.