Delta T® Heated Culture Dishes, 0.5 or 0.17 mm Coverglass, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
0420042105C 0420042105B 04200417C 04200417B
10199-772PK 193.77 CAD
10199-772 10199-774 10199-954 10199-956
Delta T® Heated Culture Dishes, 0.5 or 0.17 mm Coverglass, Bioptechs Inc.®
Culture Dishes Cell Culture Dishes
Delta T® Heated Culture Dishes are designed to simulate host conditions on the microscope stage, and provide an optimal optical environment for microscopy
  • Easily adaptable to a wide variety of specimen types; from monolayered adherent cells to brain slice and tissue preps
  • Plate, incubate, and observe without the need to transfer cells
  • Coverglass bottom for optimum optical compatibility
  • Direct heat transfer to cells through coverslip
The dishes allow users to plate cells and observe them without having to transfer them to another structure. AnMore Product Information