Jomac® Cut and Sewn Heavy Weight Terry Cloth Gloves, Loop Out, Wells Lamont
Supplier: Wells Lamont
666 766
80091-236EA 19.36 CAD
80091-236 80091-246 80091-248
Jomac® Cut and Sewn Heavy Weight Terry Cloth Gloves, Loop Out, Wells Lamont
Gloves
Jomac cut and sewn heavy weight gloves protect against up to 325 °F.
  • Natural white
  • Loop-out terry cloth
  • Knit-wrist cuff
  • Ambidextrous
  • Heavy weight
Users can clean the ambidextrous gloves in the washing machine.
Ordering information: Gloves are sold by the dozen pairs.
Caution: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Cuff style Knit wrist