Zinc(II) trifluoromethanesulfinate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Z0028-1G
Z0028-5G
TCZ0028-1GEA
130.51
CAD
TCZ0028-1G
TCZ0028-5G
Zinc(II) trifluoromethanesulfinate ≥98.0% (by titrimetric analysis)
Zinc(II) trifluoromethanesulfinate
CAS Number: 39971-65-8
MDL Number: MFCD22374174
Molecular Formula: C2F6O4S2Zn
Molecular Weight: 331.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 157
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS
Formula:
C₂F₆O₄S₂Zn Melting Pt: 157 °C |
MDL Number:
MFCD25978123 CAS Number: 39971-65-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...