Zaltoprofen ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Z0021-1G Z0021-5G
TCZ0021-1GEA 44.24 CAD
TCZ0021-1G TCZ0021-5G
Zaltoprofen ≥98.0% (by HPLC, titration analysis)
Zaltoprofen

CAS Number: 74711-43-6
MDL Number: MFCD00864323
Molecular Formula: C17H14O3S
Molecular Weight: 298.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 137


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 2-(10,11-Dihydro-10-oxodibenzo[b,f]thiepin-2-yl)propionic Acid

Formula: C₁₇H₁₄O₃S
Melting Pt: 137 °C
MDL Number: MFCD00864323
CAS Number: 74711-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1432c15d2967c40ded3002cbcf5383752f564b12&vid=1432c15d2967c40ded3002cbcf5383752f564b12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR